ContactUsLinkComponent
HLA/Terasaki Plates, Greiner Bio-One
Catalogue # 82050-706
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
undefined
HLA/Terasaki Plates, Greiner Bio-One
Catalogue # 82050-706
Fournisseur:  Greiner Bio-One
Supplier Number:  653180
Product available on GSA Advantage®
Spécifications
  • No. of wells:
    60 Wells
  • Well Volume:
    10 µL
  • Description:
    Plate with raised lid
  • Cat. no.:
    82050-706
  • Supplier No.:
    653180
Spécifications
À propos de cet article
Made of high-clarity polystyrene resin, these microtest plates are suitable for all applications for serological determination of HLA antigens, and are widely used for microbatch protein crystallization.
  • Polystyrene resin construction
  • Smooth, hydrophilic surface in most models
  • Flat-bottom design for enhanced clarity
Plates are treated to assure a smooth, hydrophilic surface for sera and cells, and come with standard raised lids or recessed lids for greater stackability. Well top is conical for localization of crystallization drop; well bottom is flat for optical clarity. Rims allow for simultaneous filling of wells. All plates have a hydrophilic tissue culture treated surface, and are nonpyrogenic, DNase-, RNase-, and human DNA-free; except for the raised 11.5 µL plates, which have untreated, hydrophobic surfaces. Plate LxWxH dimensions: 83 x 58 x 10 mm.
Frequently Bought Together