Spécifications
- No. of wells:60 Wells
- Well Volume:10 µL
- Description:Plate with raised lid
- Cat. no.:82050-706
- Supplier No.:653180
Spécifications
À propos de cet article
Made of high-clarity polystyrene resin, these microtest plates are suitable for all applications for serological determination of HLA antigens, and are widely used for microbatch protein crystallization.
- Polystyrene resin construction
- Smooth, hydrophilic surface in most models
- Flat-bottom design for enhanced clarity
Plates are treated to assure a smooth, hydrophilic surface for sera and cells, and come with standard raised lids or recessed lids for greater stackability. Well top is conical for localization of crystallization drop; well bottom is flat for optical clarity. Rims allow for simultaneous filling of wells. All plates have a hydrophilic tissue culture treated surface, and are nonpyrogenic, DNase-, RNase-, and human DNA-free; except for the raised 11.5 µL plates, which have untreated, hydrophobic surfaces. Plate LxWxH dimensions: 83 x 58 x 10 mm.