Petri Dishes and Contact Dishes, Polystyrene, Greiner bio-one
Catalog # 82050-596
Supplier: Greiner Bio-One
Product available on GSA Advantage®
Specifications
- Description:Triple Vents
- Dimensions:145W x 20H mm
- Cat. no.:82050-596
- Growth Area:141 cm2
- Supplier No.:639102
Specifications
About this item
These polystyrene petri dishes and contact dishes are available in different sizes and formats.
- Available with or without vents
- Nonsterile and sterile
- Highly transparent polystyrene construction
Manufactured from highly transparent polystyrene, dishes are heat-resistant up to 60°C for use with hot agar.
Depending on the application, vented or non-vented petri dishes and contact dishes can be selected. If applicable, sterile dishes are also available.
Besides the standard round design, a square-profile petri dish is also available. Special models include petri dishes with two or three compartments and germ count dishes.
Customers Who Bought This Also Bought