ContactUsLinkComponent
96-Well Half-Area Plate
96-Well Half Area Microplates, Greiner Bio-One
# de catalogue 82050-042
Fournisseur:  Greiner Bio-One
96-Well Half-Area Plate
96-Well Half Area Microplates, Greiner Bio-One
# de catalogue 82050-042
Fournisseur:  Greiner Bio-One
Spécifications
  • Format:
    LUMITRAC™ 200
  • Sterility:
    Non sterile
  • Color:
    White
  • Surface Treatment:
    Medium binding
  • Bottom Color:
    White
  • Supplier No.:
    675075
Spécifications
A propos de cet article
Polystyrene half area microplates are compatible with manual and automated pipetting, and dispensing and reading devices used with standard 96-well plates.
  • Polystyrene construction
  • Chimney-well design helps reduce cross-contamination
  • Clear film bottoms
  • Suitable for cell-based assays
This design allows up to a 50% sample and reagent reduction and the flat bottoms allow stackability with or without lids
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardized path-length, eliminating the need for path-length determinations. MICROLON™ formats are ideal for consistent performance in binding assays. White LUMITRAC™ plates are ideal for luminescence assays, while black FLUOTRAC™ plates are suitable for fluorescence assays. Both formats are also suitable for cell-based assays.
The chimney style well design reduces the risk of cross-contamination and aids sealing by cap mat, thermal sealer, or adhesive sealer.
Customers Who Bought This Also Bought