ContactUsLinkComponent
Manual Serological Acrylic Pipette Rack
Serological Pipette Rack, IBI Scientific
# de catalogue 82031-544
Fournisseur:  IBI Scientific
Manual Serological Acrylic Pipette Rack
Serological Pipette Rack, IBI Scientific
# de catalogue 82031-544
Fournisseur:  IBI Scientific
Spécifications
  • Dimensions:
    9.4W x 28.6D x 40.3H cm (311/16 x 111/4 x 157/8")
  • Description:
    Small Pipette Rack
  • Cat. no.:
    82031-544
  • Supplier No.:
    PR-40
Spécifications
A propos de cet article
These stands provide storage space for serological pipettes
Constructed of heavy-duty 6mm (1/4") clear-cast acrylic with orange shelves. Angled shelves are slightly indented for better access. Solid bottom and rubber feet for stability.
Customers Who Bought This Also Bought