Nalgene® Biotainer® Polycarbonate Carboys, Thermo Scientific
Product Details & Documents
Sterile carboys are molded from blue-tinted polycarbonate and provide safe storage for contents at temperatures from −85 °C (−121 °F) to near boiling
- Space-saving square shape
Carboys meet USP-VI and are noncytotoxic and nonpyrogenic. Gamma irradiated to a sterility assurance level of 10⁻⁶.
Ideal for storage and transport of media, sera, antigens, and other biological reagents. Carboys feature molded graduations in liters and gallons and indented, ribbed handgrips.
Ordering information: Each carboy is supplied with a 48mm screw cap with a silicone liner. Individually bagged.
Spécifications
- Capacité:10 L (2.6 gal.)
- Désignation:Biotainer® Carboy with Handle
- Cat. No.:82028-474
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



