ContactUsLinkComponent
VWR® Acrylic Pipettor Stands
Catalogue # 82024-542
Fournisseur:  VWR International
undefined
VWR® Acrylic Pipettor Stands
Catalogue # 82024-542
Fournisseur:  VWR International
Spécifications
  • Description:
    4-Place Stand
  • Cat. no.:
    82024-542
Spécifications
À propos de cet article
Convenient three-, four-, and six-place stands are designed to hold most major brands of pipettors.
  • Clear acrylic construction
  • Designed to hold most major brands of pipettors
They are constructed of clear acrylic and accommodate a variety of laboratory applications and needs. Upper slots are 25mm (1") wide and lower slots are 20mm (13/16") wide.
Frequently Bought Together