VWR® Acrylic Pipettor Stands
Catalogue # 82024-542
Fournisseur: VWR International
Spécifications
- Description:4-Place Stand
- Cat. no.:82024-542
Spécifications
À propos de cet article
Convenient three-, four-, and six-place stands are designed to hold most major brands of pipettors.
- Clear acrylic construction
- Designed to hold most major brands of pipettors
They are constructed of clear acrylic and accommodate a variety of laboratory applications and needs. Upper slots are 25mm (1") wide and lower slots are 20mm (13/16") wide.
Frequently Bought Together