Order Entry
ContactUsLinkComponent
VWR** Freezer Racks
VWR® Freezer Racks
# de catalogue 82024-520
Fournisseur:  VWR International
VWR** Freezer Racks
VWR® Freezer Racks
# de catalogue 82024-520
Fournisseur:  VWR International

Spécifications

  • Description:
    Four-Shelf Rack, 2" Boxes
  • Cat. no.:
    82024-520
  • L×D×H:
    21.8 x 5.5 x 8.9"

Spécifications

A propos de cet article

Corrosion-resistant, stainless steel horizontal racks feature handles for easy transport.

  • Corrosion-resistant
  • Stainless steel fabrication
  • Multiple shelf counts

Three-shelf rack fits in upright freezers and holds one 7.6 cm (3") box per shelf. Four-shelf rack fits in chest freezers and holds one 5.1 cm (2") box per shelf.

Customers Who Bought This Also Bought