VWR® 8-Well Centrifuge Tube Rack
# de catalogue 82024-452
Fournisseur: VWR International
Spécifications
- Cat. no.:82024-452
- W×D×H:14 x 7.7 x 8.8 cm (51/2 x 3 x 31/2")
Spécifications
A propos de cet article
Rack is ideal for holding up to eight 50 mL conical centrifuge tubes and features a slotted bottom for secure handling.
- Autoclavable
- Resistant to chemicals
Rack can also accommodate other types and sizes of tubes. Tubes sit in 30mm square holes. Epoxy-coated steel wire rack is chemical-resistant and autoclavable.
Customers Who Bought This Also Bought