ContactUsLinkComponent
VWR* Pipettor Stand
VWR® Pipettor Stand
# de catalogue 82010-756
Fournisseur:  VWR International
VWR* Pipettor Stand
VWR® Pipettor Stand
# de catalogue 82010-756
Fournisseur:  VWR International
Spécifications
  • Color:
    Clear with Blue Lower Rack
  • Cat. no.:
    82010-756
Spécifications
A propos de cet article
Acrylic, four-place stand provides efficient, convenient storage for most major brands of pipettors.
  • Acrylic construction
  • Holds up to four pipettors
  • Efficient, convenient storage
Upper slots are 25mm wide. Lower slots are 20mm wide.
Customers Who Bought This Also Bought