Absorbent Liners, Jaece
# de catalogue 82002-608
Fournisseur: Jaece
Spécifications
- Description:50.8 cm x 91.4 m (20" x 300') Roll
- Cat. no.:82002-608
- Supplier No.:AL20600
Spécifications
A propos de cet article
Made of highly absorbent three-ply tissue with a waterproof polypropylene backing that is resistant to most chemicals.
- Cushions surfaces while absorbing liquids
- Can be cut to size
Keeps spills from spreading and protects benchtops, floors, and carts.
Ordering information: Liner 82002-606 is packaged one roll per case.
Customers Who Bought This Also Bought