ContactUsLinkComponent
RACK 96 PCR FLUORESCENT YELLOW CS 25
Axygen® PCR Tube Storage Racks, Corning
# de catalogue 80086-082
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
RACK 96 PCR FLUORESCENT YELLOW CS 25
Axygen® PCR Tube Storage Racks, Corning
# de catalogue 80086-082
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Spécifications
  • Color:
    Fluorescent Yellow
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Cat. no.:
    80086-082
  • Supplier No.:
    R-96-PCR-FY
Spécifications
A propos de cet article
PP with lid.
  • Fluorescent color base with translucent lid
  • Durable, chemically inert, and can withstand extended duration at sub-zero temperatures
Designed to hold 0.2, and 0.5 ml PCR tubes.
Ordering information: Assorted pack includes one each of blue, green, violet, red and yellow.
Customers Who Bought This Also Bought