Axygen® PCR Tube Storage Racks, Corning
# de catalogue 80086-082
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Color:Fluorescent Yellow
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:80086-082
- Supplier No.:R-96-PCR-FY
Spécifications
A propos de cet article
PP with lid.
- Fluorescent color base with translucent lid
- Durable, chemically inert, and can withstand extended duration at sub-zero temperatures
Designed to hold 0.2, and 0.5 ml PCR tubes.
Ordering information: Assorted pack includes one each of blue, green, violet, red and yellow.
Customers Who Bought This Also Bought