Erlenmeyer Flasks with [ST] Joint, Chemglass
# de catalogue 80070-042
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Capacity:6000 ml
- ST Joint No.:24/40
- Cat. no.:80070-042
- Supplier No.:CG-1542-23
Spécifications
A propos de cet article
These Erlenmeyer Flasks have a standard taper outer joint.
Customers Who Bought This Also Bought