ContactUsLinkComponent
FLASK, ERLENMEYER, OUTER JOINT
Erlenmeyer Flasks with [ST] Joint, Chemglass
# de catalogue 80070-026
Fournisseur:  Chemglass
Small Business Enterprise
FLASK, ERLENMEYER, OUTER JOINT
Erlenmeyer Flasks with [ST] Joint, Chemglass
# de catalogue 80070-026
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Capacity:
    2000 ml
  • ST Joint No.:
    24/40
  • Cat. no.:
    80070-026
  • Supplier No.:
    CG-1542-15
Spécifications
A propos de cet article
These Erlenmeyer Flasks have a standard taper outer joint.
Customers Who Bought This Also Bought