ContactUsLinkComponent
FILTER FLASK, REPL. HOSE, 15L
DURAN® Filtering Flask, with Replaceable Hose Connection, Chemglass
# de catalogue 80061-434
Fournisseur:  Chemglass
Small Business Enterprise
FILTER FLASK, REPL. HOSE, 15L
DURAN® Filtering Flask, with Replaceable Hose Connection, Chemglass
# de catalogue 80061-434
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Capacity:
    15 L
  • Description:
    Heavy wall flask, with replaceable hose connection
  • Cat. no.:
    80061-434
  • Supplier No.:
    CG-1560-08
Spécifications
A propos de cet article
Duran® heavy wall filtering flask with a unique removable PPN hose connection.
  • Hose connection is secured to the flask via a silicone rubber seal
  • Bottle shaped filter flask, uncoated or plastic coated with removable PPN hose connection
BodyTextAccessories: Replacement PPN hose connection set contains PPN tubulation, Silicone rubber seal, Male PPN hose connection and Female PPN connection.
Customers Who Bought This Also Bought