Nalgene® Disposable Erlenmeyer Flasks with Vented Closure, PETG, Sterile, Plain Bottom, Thermo Scientific
# de catalogue 74910-008
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:125 ml
- Cat. no.:74910-008
- Thread:38 - 430
- Supplier No.:4115-0125
Spécifications
A propos de cet article
Made of lightweight, clear PETG plastic, these flasks are ideal for shaker and suspension cell culture, as well as media preparation or storage.
- Flasks feature molded-in graduations
- Non-pyrogenic, non-cytotoxic
- Sterilized to SAL 10-6
The high-density polyethylene closure has a hydrophobic 0.2µm PTFE membrane that allows sterile air exchange. Sterile disposable flasks reduce the chance of cross contamination.
Ordering information: Individually packaged.
Customers Who Bought This Also Bought