Nalgene® Square Media Bottles, PET, Sterile, Graduated, Thermo Scientific
Catalogue # 73520-442
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:125 ml (4.2 oz.)
- Description:With closure
- Cat. no.:73520-442
- Brim Capacity:184 ml (6.2 oz.)
- Supplier No.:342040-0125
Spécifications
À propos de cet article
Bottles are lightweight, shatter-resistant, and provide excellent gas barrier properties for storage and shipping of liquid media, buffers, and sera.
- Noncytotoxic
Sterile and nonpyrogenic to save time and prevent costly cleaning preparation. Bottles feature molded-in graduations and molded-in neck ring for use with tamper-evident shrink sleeves. Suitable for air shipment and tested to ensure leakproof performance.
Certifications: Bottles meet USP Class VI, USP 661, and European Pharmacopeia for Abnormal Toxicity standards.
Ordering information: Packaged in shrink-wrapped trays. Trays are double-bagged for critical environment use. Bottles are packed 24 per tray, except bottles 16010-104 and 16010-106, which are packed 12 per tray. All bottles feature a 38-430 neck finish; a high-density polyethylene closure is included where indicated.
Frequently Bought Together