Order Entry
ContactUsLinkComponent
Nalgene High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
Nalgene® High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
# de catalogue 73520-068
Nalgene High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
Nalgene® High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
# de catalogue 73520-068

Spécifications

  • Cap Size:
    20-415
  • Capacity:
    60 ml (2 oz.)
  • Cat. no.:
    73520-068
  • Supplier No.:
    342089-0002

Spécifications

A propos de cet article

HDPE bottles feature a narrow mouth and leakproof closure design.

Manufactured and packaged in controlled environments to minimize biological and particulate contamination. Gamma irradiated to 10⁻⁶ sterility assurance levels.

Certifications: Bottles meet USP-VI and are noncytotoxic and nonpyrogenic.
Ordering information: Supplied with white polypropylene closures.
Customers Who Bought This Also Bought