Lab Mailers and Mailing Sleeves for Specimen Transport, Sonoco ThermoSafe
Catalog # 73320-084
Supplier: Sonoco Thermosafe
Specifications
- Description:5 tube specimen mailer, 105×13 mm
- Dimensions:10.5×1.3 cm (41/8×1/8")
- Cat. no.:73320-084
- Tube Dimensions:105×13 mm
- Supplier No.:339
Specifications
About this item
Lightweight, economical EPS shippers provide maximum breakage protection for known non-hazardous materials during specimen transport.
- For VacuTainer®, Microtainer®, and other collection/transport tubes
- Available in multi-tube, slide, and tube/slide combinations
- Lightweight polystyrene foam saves postage
- Reusable and recyclable
A unique array of specimen shippers fills the need for protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.
Frequently Bought Together