ContactUsLinkComponent
Automation Pipette Tips for Tecan Genesis and Freedom EVO Workstations, Thermo Scientific
Catalogue # 72830-324
   Low Manufacturing Impact
Automation Pipette Tips for Tecan Genesis and Freedom EVO Workstations, Thermo Scientific
Catalogue # 72830-324
Supplier Number:  903-252G
   Low Manufacturing Impact
Spécifications
  • Volume:
    200 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Packed:
    96 Tips/Tray, 10 Trays/Case
  • Cat. no.:
    72830-324
  • Tip Style:
    Wide bore
  • For Use With:
    Tecan automated workstations
  • Supplier No.:
    903-252G
Spécifications
À propos de cet article
Thermo Scientific provides a solution for your Tecan robotic pipette tips.
These automation pipette tips have been designed, validated, and tested on a Tecan Genesis robot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our Tecan workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Frequently Bought Together