Automation Pipette Tips for Tecan Genesis and Freedom EVO Workstations, Thermo Scientific
Catalogue # 72830-324
Fournisseur: Thermo Fisher Scientific
Spécifications
- Volume:200 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Packed:96 Tips/Tray, 10 Trays/Case
- Cat. no.:72830-324
- Tip Style:Wide bore
- For Use With:Tecan automated workstations
- Supplier No.:903-252G
Spécifications
À propos de cet article
Thermo Scientific provides a solution for your Tecan robotic pipette tips.
These automation pipette tips have been designed, validated, and tested on a Tecan Genesis robot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our Tecan workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Frequently Bought Together