BioRobotix® Pipette Tips for Rosys® and QIAGEN® Workstations, Molecular BioProducts
# de catalogue 72830-252
Fournisseur: Thermo Scientific
Spécifications
- Volume:300 µl
- Sterile:+
- Tip Style:Clear
- Supplier No.:951-261
Spécifications
A propos de cet article
Compatible with Rosys® and QIAGEN® style workstations.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as liquid sensing tips to maximize the workstation abilites, or clear polypropylene for applications that do not require liquid sensing. Three volume sizes include 250, 300 and 1100 µl. Filter tips utilize the ART® barrier.
Ordering information: Tips are packaged 96 per tray, 10 trays per case.
Customers Who Bought This Also Bought