ContactUsLinkComponent
BioRobotix® Pipette Tips for Rosys® and QIAGEN® Workstations, Molecular BioProducts
Catalogue # 72830-248
Fournisseur:  Thermo Scientific
   Low Manufacturing Impact
BioRobotix® Pipette Tips for Rosys® and QIAGEN® Workstations, Molecular BioProducts
Catalogue # 72830-248
Fournisseur:  Thermo Scientific
Supplier Number:  951-251
   Low Manufacturing Impact
Spécifications
  • Volume:
    300 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Cat. no.:
    72830-248
  • Sterile:
    +
  • Tip Style:
    Liquid Sensing
  • Supplier No.:
    951-251
Spécifications
À propos de cet article
Compatible with Rosys® and QIAGEN® style workstations.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as liquid sensing tips to maximize the workstation abilites, or clear polypropylene for applications that do not require liquid sensing. Three volume sizes include 250, 300 and 1100 µl. Filter tips utilize the ART® barrier.
Ordering information: Tips are packaged 96 per tray, 10 trays per case.
Frequently Bought Together