BioRobotix® Pipette Tips for Tecan® and Cavro® Workstations, Molecular BioProducts
Catalogue # 72830-178
Fournisseur: Thermo Fisher Scientific
Spécifications
- Volume:1000 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Packed:96/Tray, 10 trays
- Cat. no.:72830-178
- Tip Style:Liquid Sensing
- Supplier No.:904-252
Spécifications
À propos de cet article
Compatible with Tecan®, Cavro®, and Packard MultiPROBE II HT style workstations.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as liquid sensing tips to maximize the workstation abilites, or clear polypropylene for applications that do not require liquid sensing. Total of six volumes available, for both tip styles, for increased repeatability in any application. Filter tips utilize the ART® barrier for increased protection against contamination.
Frequently Bought Together