ContactUsLinkComponent
Tubing for Microlab* 500 Series Dispensers/Diluters
Syringes and Tubing for MICROLAB® 500 Series Dispensers/Diluters, Hamilton
# de catalogue 72310-026
Fournisseur:  Hamilton
Syringes and Tubing for MICROLAB® 500 Series Dispensers/Diluters, Hamilton
# de catalogue 72310-026
Fournisseur:  Hamilton
Spécifications
  • Cat. no.:
    72310-026
  • Gauge:
    18
  • Supplier No.:
    240130
Spécifications
A propos de cet article
For use with 25227-830 series dispensers/diluters.
Note: For dual dispenser models, use reagent (left side) syringes on both sides.
Customers Who Bought This Also Bought