PYREX® 47 mm Microfiltration Glassware Apparatus, Corning
# de catalogue 71001-592
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Description:PYREX® 47 mm Microfiltration Glassware Apparatus - Complete
- Environmentally Preferable:Low Manufacturing Impact,Waste Reducing
- Cat. no.:71001-592
- Diameter ×H:230 x 75 mm
- Funnel Capacity:300 ml
- Supplier No.:33970-47
- Environmental certification:I-RECs, ISO 14001 - Monterrey
Spécifications
A propos de cet article
These microfiltration components are designed for the filtration of aqueous liquids. They may be used for the filtration of cell culture media, buffers or for particulate analysis of water samples.
- Units have coarse porosity frits and accommodate 47 mm filters
- All-glass design ensures chemical compatibility with most organic solvents
- Components are interchangeable with competitive products
Ordering information: Heavy wall filtering flasks are also available on request (sold separately).
Delivery: The unit includes 300 ml funnel, 47 mm fritted glass support base, aluminum clamp and #8 silicone stopper.
Customers Who Bought This Also Bought