Closures for Storage/Reaction Vials with Closure, Chemglass
# de catalogue 71000-556
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:SURE-LINK™ Cap, Open Top, 24-414, polypropylene, green, TFE septa
- Cat. no.:71000-556
- Supplier No.:CG-4911-20