Tissue Homogenizer, Dounce, Ace Glass Incorporated
# de catalogue 71000-514
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Capacity:1 ml
- Description:Dounce Tissue Homogenizer with Two Glass Pestles
- Cat. no.:71000-514
- O.D.×H:11 x 48 mm
- Overall Length:125 mm
- Supplier No.:8343-01
Spécifications
A propos de cet article
This Dounce tissue homogenizer is supplied with two pestles, one "loose" and one "tight", to ensure dissociation of cells into fine particles with minimal damage to cell nuclei
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
Use the large-clearance pestle for initial reduction of soft tissue. Complete the homogenization with the small-clearance pestle. Particularly useful in enzyme studies where heat build-up must be avoided. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought