ContactUsLinkComponent
13Mm Lyophilization Stopper
Lyophilization-Style Stoppers, Rubber, Wheaton
# de catalogue 71000-060
Small Business Enterprise
13Mm Lyophilization Stopper
Lyophilization-Style Stoppers, Rubber, Wheaton
# de catalogue 71000-060
Small Business Enterprise
Spécifications
  • Color:
    Gray
  • Description:
    Stopper, 2-leg lyophilization
  • Material:
    Chlorobutyl
  • Fits:
    Mouth with ID×OD 7×13 mm
  • Supplier No.:
    W224100-093
Spécifications
A propos de cet article
Wheaton Lyophilization Stoppers are manufactured from the highest quality raw materials and are manufactured to yield exceptional dimensional stability
Stoppers are autoclavable.
Customers Who Bought This Also Bought