Order Entry
ContactUsLinkComponent
Universal Handle
Stainless Steel Quick Disconnect Handles, Micronova
Catalog # 68400-164
Supplier:  Micronova
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Universal Handle
Stainless Steel Quick Disconnect Handles, Micronova
Catalog # 68400-164
Supplier:  Micronova
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Stainless steel quick disconnect extendable handle
  • Length:
    Extendable from 96.5 - 182.9 cm (38 - 72")
  • Material:
    Stainless steel 304, electropolished
  • Cat. no.:
    68400-164
  • Supplier No.:
    SSU-1

Specifications

About this item

Universal electropolished stainless steel handles with a quick disconnect connection that works with the entire range of Micronova adapters, tools and mop heads for simplified and versatile cleaning.

  • Universal handles work with all Quick Disconnect tools and mop adapters
  • 304 electropolished stainless steel with polyketone components
  • Strong, hard-wearing connections for long lasting use
  • Available in a range of fixed or extendable lengths from 8 in (20 cm) to 10 ft (3 m)
  • Quick Disconnect end attaches directly to QD mops, tools and adapters
  • SSA-3 adapter connects QD handles to plastic threaded mops, tools and adapters
  • Use the Adjustable Angle Adapters to reach over, under and around obstacles in the cleanroom
  • Good compatibility with disinfectants
  • Autoclavable

The Micronova Stainless Steel Universal Handles attach directly to the stainless steel Quick Disconnect (QD series) adapters and tools. Use the SSA-3 Quick Disconnect Adapter to attach to the plastic ACME threaded (QP series) adapters and tools. The Adjustable Angle Adapters can be used to create an adjustable angled shape between a handle and mop adapter to clean above and below fixtures, as well as, inside isolators and containment hoods.

Customers Who Bought This Also Bought