Spécifications
- Description:Hamilton Model 1025 Replacement Plunger
- Cat. no.:66513-626
- Supplier No.:13271
- Termination:LT, LTN, TLL
Spécifications
A propos de cet article
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
These replacement plungers are for Hamilton 1000 series Gastight® syringes ranging in volume from 1 to 2 ml.
Each is manufactured to achieve the highest level of accuracy and precision possible.