Nessler Tube Supports, Bennett Wood
# de catalogue 66176-722
Fournisseur: Bennett Wood Specialties
Spécifications
- Description:Holds 50 mL Tall Form Nessler Tubes
- Cat. no.:66176-722
- Supplier No.:6765-50
Spécifications
A propos de cet article
Holds 12 Nessler tubes
Made of dull, black wood, with white opaque reflector set at an angle below tubes. Tubes are supported by a ledge to prevent their bottoms from coming in contact with reflector. Supplied without tubes.
Customers Who Bought This Also Bought