To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Size:1 L bottle
- Cat. no.:66115-140
- For Use With:Non-Ratio Meters
- Supplier No.:85034
Spécifications
A propos de cet article
EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers
EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers
One year shelf life.
Ready to use at their stated value without mixing or diluting. Composed of submicron solid polymer beads suspended in ultra-pure water. Guaranteed accuracy within 1%. Safe, non-toxic, non-carcinogenic alternative to Formazin. Available for non-ratio and ratio meters.