Shallow Form Watch Glasses, Walter Stern
# de catalogue 66112-060
Fournisseur: Walter Stern
Spécifications
- Ø:50 mm (2")
- Description:Watch glass
- Cat. no.:66112-060
- Supplier No.:110-102
Spécifications
A propos de cet article
Concave, clear, annealed glass covers have fire-polished edges
Used to return condensation to beakers. Manufactured from soda lime glass.
Customers Who Bought This Also Bought