ContactUsLinkComponent
Manual Decrimpers For All Crimp Seals, 32 mm
Crimpers, Decrimpers and Decapping Pliers, Thermo Scientific
# de catalogue 66064-374
Fournisseur:  Thermo Scientific
Manual Decrimpers For All Crimp Seals, 32 mm
Crimpers, Decrimpers and Decapping Pliers, Thermo Scientific
# de catalogue 66064-374
Fournisseur:  Thermo Scientific
Spécifications
  • Description:
    Decrimper, For Crimp Seals
  • Cat. no.:
    66064-374
  • Seal Size:
    32 mm
  • Supplier No.:
    C4032-102
Spécifications
A propos de cet article
Tools are designed to manually secure or release standard crimp seals to and from vials.
Decrimper is a user-friendly method of removing crimp seals and is recommended if vials are to be reused. If vials are not to be reused, the decapping pliers are a more economical option.
Crimper is adjustable to accommodate various septa thicknesses.
Customers Who Bought This Also Bought