Crimpers, Decrimpers and Decapping Pliers, Thermo Scientific
# de catalogue 66064-374
Fournisseur: Thermo Scientific
Spécifications
- Description:Decrimper, For Crimp Seals
- Cat. no.:66064-374
- Seal Size:32 mm
- Supplier No.:C4032-102
Spécifications
A propos de cet article
Tools are designed to manually secure or release standard crimp seals to and from vials.
Decrimper is a user-friendly method of removing crimp seals and is recommended if vials are to be reused. If vials are not to be reused, the decapping pliers are a more economical option.
Crimper is adjustable to accommodate various septa thicknesses.
Customers Who Bought This Also Bought