Crimpers, Decrimpers and Decapping Pliers, Thermo Scientific
# de catalogue 66064-270
Fournisseur: Thermo Scientific
Spécifications
- Description:Crimper, For Flip-Off Seals
- Cat. no.:66064-270
- Seal Size:13 mm
- Supplier No.:C4013-100F
Spécifications
A propos de cet article
Tools are designed to manually secure or release standard crimp seals to and from vials.
Decrimper is a user-friendly method of removing crimp seals and is recommended if vials are to be reused. If vials are not to be reused, the decapping pliers are a more economical option.
Crimper is adjustable to accommodate various septa thicknesses.
Customers Who Bought This Also Bought