ContactUsLinkComponent
Manual Crimpers for Flip-Off Crimp Seals, 13 mm
Crimpers, Decrimpers and Decapping Pliers, Thermo Scientific
# de catalogue 66064-270
Fournisseur:  Thermo Scientific
Manual Crimpers for Flip-Off Crimp Seals, 13 mm
Crimpers, Decrimpers and Decapping Pliers, Thermo Scientific
# de catalogue 66064-270
Fournisseur:  Thermo Scientific
Spécifications
  • Description:
    Crimper, For Flip-Off Seals
  • Cat. no.:
    66064-270
  • Seal Size:
    13 mm
  • Supplier No.:
    C4013-100F
Spécifications
A propos de cet article
Tools are designed to manually secure or release standard crimp seals to and from vials.
Decrimper is a user-friendly method of removing crimp seals and is recommended if vials are to be reused. If vials are not to be reused, the decapping pliers are a more economical option.
Crimper is adjustable to accommodate various septa thicknesses.
Customers Who Bought This Also Bought