Seals for 20 mm Headspace Vials, Thermo Scientific
# de catalogue 66030-802
Fournisseur: Thermo Scientific
Spécifications
- Septum:None
- Description:Seals, Tear-Off
- Cat. no.:66030-802
- Seal Type:Aluminum
- Supplier No.:C4020-5A
Spécifications
A propos de cet article
Seals have attached septa, unless otherwise specified.
Customers Who Bought This Also Bought