ContactUsLinkComponent
White Plastic Screw Cap
White Plastic Screw Caps, WHEATON®
Catalog # 66021-861
Small Business Enterprise
White Plastic Screw Cap
White Plastic Screw Caps, WHEATON®
Catalog # 66021-861
Small Business Enterprise
Specifications
  • Cap size:
    22-400
  • Cap Type:
    PE cap
  • Cat. no.:
    66021-861
  • Liner Material:
    Unlined
  • Supplier No.:
    241317
Specifications
About this item
White plastic screw caps fit Wheaton vials (see 66021-679 series) and other glass scintillation vials.
  • Suitable for marking
  • Not autoclavable
  • Available unlined or choose from a variety of liners
Customers Who Bought This Also Bought