Serum Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
# de catalogue 66013-122
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap included:No
- Cap Size:13 mm
- Capacity:3.50 ml
- Color:Clear
- Description:Serum vials
- Cat. no.:66013-122
- Diameter ×H:18×45 mm
- Supplier No.:62113D-312
Spécifications
A propos de cet article
Ideally suited for handling, containing, and storing a variety of liquids, including reagents, vaccines, and chromatography samples.
- Autoclavable
These crimp-top serum vials are lighter in weight than molded bottles. More closely held tolerances and uniformity of glass allow the vials to be lyophilized.
Ordering information: Vials available in clear or amber glass, without closure. Stoppers and seals sold separately.
Customers Who Bought This Also Bought