Order Entry
ContactUsLinkComponent
Shorty Vial*
Shorty Vials®, Borosilicate Glass, Wheaton, DWK Life Sciences
# de catalogue 66011-868
Small Business Enterprise
Shorty Vial*
Shorty Vials®, Borosilicate Glass, Wheaton, DWK Life Sciences
# de catalogue 66011-868
Small Business Enterprise

Spécifications

  • Cap included:
    No
  • Cap Size:
    13-425
  • Capacity:
    2.00 ml
  • Color:
    Clear
  • Description:
    Vials Without Caps
  • Diameter ×H:
    15×28 mm
  • Supplier No.:
    224821

Spécifications

A propos de cet article

These shorty vials have a low profile for greater stability and reduced dead volume above the sample.

  • Made from low extractable borosilicate glass
  • Superior chemical resistance

Vials are available with or without caps and a choice of liners. PTFE faced rubber lined caps provide an inert and chemically-resistant barrier for harsh, sensitive, and reactive samples. Caps with 14B rubber liner are suitable for aqueous samples. Vials and caps are autoclavable.

Certifications: Vials meet ASTM E438 Type 1, Class A and USP Type 1 standards.
Ordering information: Vials are packaged in Lab File® partitioned, corrugated trays, then trays are shrink-wrapped to reduce particulate contamination.
Customers Who Bought This Also Bought