ContactUsLinkComponent
PTFE/Silicone Rubber Septa
PTFE-Faced Silicone Rubber Septa, Autoclavable, DWK Life Sciences
# de catalogue 66010-796
PTFE-Faced Silicone Rubber Septa, Autoclavable, DWK Life Sciences
# de catalogue 66010-796
Spécifications
  • Ø:
    20 mm
  • Description:
    PTFE-faced silicone rubber septa
  • Thickness:
    0.005" PTFE, 0.060" silicone rubber
  • Cat. no.:
    66010-796
  • Supplier No.:
    73818-20
Spécifications
A propos de cet article
Ideal for use with open-top caps to access container contents with a syringe.
  • Resists coring
  • Highly chemically-resistant PTFE facing maintains integrity of content
  • Silicone rubber backing allows repeated puncturing through the seal
  • Recommended for use in all autosamplers using screw thread vials
Customers Who Bought This Also Bought