Polypropylene Screw Caps, Linerless, Kimble Chase
# de catalogue 66010-680
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Size:15-415
- Color:Natural
- Supplier No.:73805B-15415
Spécifications
A propos de cet article
Polypropylene caps feature one-piece construction with a seal achieved by a unique interior design.
Caps withstand repeated autoclaving when loosened or removed from tubes.
Certifications: Designed from ASTM Specification E982
Customers Who Bought This Also Bought