ContactUsLinkComponent
Caps closures for Scintillation Vial White Polypropylene
Scintillation Vial Closures, Kimble Chase
# de catalogue 66009-256
Caps closures for Scintillation Vial White Polypropylene
Scintillation Vial Closures, Kimble Chase
# de catalogue 66009-256
Spécifications
  • Color:
    White
  • Description:
    Polypropylene Closures
  • Cat. no.:
    66009-256
  • Liner Material:
    Pulp-Backed Aluminum Foil
  • Thread Size:
    15-425
  • Supplier No.:
    74521-15425
Spécifications
A propos de cet article
Suitable for use with various strong oxidizing agent mixtures.
  • Top is suitable for marking
  • Ideal for scintillation vials
Customers Who Bought This Also Bought