Scintillation Vial Closures, Kimble Chase
# de catalogue 66009-256
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Color:White
- Description:Polypropylene Closures
- Cat. no.:66009-256
- Liner Material:Pulp-Backed Aluminum Foil
- Thread Size:15-425
- Supplier No.:74521-15425
Spécifications
A propos de cet article
Suitable for use with various strong oxidizing agent mixtures.
- Top is suitable for marking
- Ideal for scintillation vials
Customers Who Bought This Also Bought