Septa and Seals for Headspace Vials, SUN SRI™
# de catalogue 66008-038
Fournisseur: Thermo Scientific
Spécifications
- Septum:Natural PTFE/Natural Silicone
- Description:Septa, 20 mm
- Thickness:2.8 mm (0.110")
- Cat. no.:66008-038
- Supplier No.:200 962
Spécifications
A propos de cet article
Customers Who Bought This Also Bought