Versilon™ C-219-A Flexible Chemical Transfer Tubing
Catalog # 63009-404
Supplier: Saint Gobain Life Sciences
Specifications
- Tubing ext.Ø:½"
- Tubing I.D.:⅜"
- Wall thickness:¹⁄₁₆"
- Durometer hardness:Shore A, 67
- Cat. no.:63009-404
- Supplier No.:ABH02027
Specifications
About this item
General use clear vinyl tubing is supplied in 30.5 m (100') lengths.
Customers Who Bought This Also Bought