ContactUsLinkComponent
TUBING VINCON 3/8X1/2 100FT
Versilon™ C-219-A Flexible Chemical Transfer Tubing
Catalog # 63009-404
TUBING VINCON 3/8X1/2 100FT
Versilon™ C-219-A Flexible Chemical Transfer Tubing
Catalog # 63009-404
Specifications
  • Tubing ext.Ø:
    ½"
  • Tubing I.D.:
    ⅜"
  • Wall thickness:
    ¹⁄₁₆"
  • Durometer hardness:
    Shore A, 67
  • Cat. no.:
    63009-404
  • Supplier No.:
    ABH02027
Specifications
About this item
General use clear vinyl tubing is supplied in 30.5 m (100') lengths.
Customers Who Bought This Also Bought