ContactUsLinkComponent
TUBING VINCON 1/2 X 3/4 100FT
Versilon™ C-219-A Flexible Chemical Transfer Tubing
# de catalogue 63009-365
TUBING VINCON 1/2 X 3/4 100FT
Versilon™ C-219-A Flexible Chemical Transfer Tubing
# de catalogue 63009-365
Spécifications
  • Tubing I.D.:
    ½"
  • Tubing O.D.:
    ¾"
  • Wall thickness:
    ⅛"
  • Durometer hardness:
    Shore A, 67
  • Supplier No.:
    ABH02038
Spécifications
A propos de cet article
General use clear vinyl tubing is supplied in 30.5 m (100') lengths.
Customers Who Bought This Also Bought