Versilon™ C-219-A Flexible Chemical Transfer Tubing
# de catalogue 63009-365
Fournisseur: Saint Gobain Life Sciences
Spécifications
- Tubing I.D.:½"
- Tubing O.D.:¾"
- Wall thickness:⅛"
- Durometer hardness:Shore A, 67
- Supplier No.:ABH02038
Spécifications
A propos de cet article
General use clear vinyl tubing is supplied in 30.5 m (100') lengths.
Customers Who Bought This Also Bought