Versilon™ C-219-A Flexible Chemical Transfer Tubing
# de catalogue 63009-353
Fournisseur: Saint Gobain Life Sciences
Spécifications
- Tubing I.D.:⁵⁄₁₆"
- Tubing O.D.:⁷⁄₁₆"
- Wall thickness:¹⁄₁₆"
- Durometer hardness:Shore A, 67
- Supplier No.:ABH02022
Spécifications
A propos de cet article
General use clear vinyl tubing is supplied in 30.5 m (100') lengths.
Customers Who Bought This Also Bought