ContactUsLinkComponent
VWR* Amber Latex Rubber Tubing
VWR® Amber Latex Rubber Tubing
# de catalogue 62996-509
Fournisseur:  VWR International
VWR* Amber Latex Rubber Tubing
VWR® Amber Latex Rubber Tubing
# de catalogue 62996-509
Fournisseur:  VWR International
Spécifications
  • Tubing I.D.:
    ¹/₄"
  • Tubing O.D.:
    ⁷/₁₆"
  • Wall thickness:
    ³/₃₂"
  • Description:
    50' Reel
  • Cat. no.:
    62996-509
Spécifications
A propos de cet article
High-quality rubber tubing recommended for all kinds of glass connections where highly elastic, clinging tubing is desired.
  • Durometer hardness: Shore A, 35
  • Tensile strength: 3500 psi
  • Translucent and durable
Amber latex rubber tubing is available in 50' lengths on reels or in boxes.
Ordering information: This product comes in three pieces.
Customers Who Bought This Also Bought