VWR® Amber Latex Rubber Tubing
# de catalogue 62996-473
Fournisseur: VWR International
Spécifications
- Tubing I.D.:¹/₄"
- Tubing O.D.:³/₈"
- Wall thickness:¹/₁₆"
- Description:50' Reel
- Cat. no.:62996-473
Spécifications
A propos de cet article
High-quality rubber tubing recommended for all kinds of glass connections where highly elastic, clinging tubing is desired.
- Durometer hardness: Shore A, 35
- Tensile strength: 3500 psi
- Translucent and durable
Amber latex rubber tubing is available in 50' lengths on reels or in boxes.
Ordering information: This product comes in three pieces.
Customers Who Bought This Also Bought