ContactUsLinkComponent
VWR* Black Latex Rubber Tubing
VWR® Latex Rubber Tubing, Black
# de catalogue 62995-721
Fournisseur:  VWR International
VWR* Black Latex Rubber Tubing
VWR® Latex Rubber Tubing, Black
# de catalogue 62995-721
Fournisseur:  VWR International
Spécifications
  • Tubing I.D.:
    ⅜"
  • Tubing O.D.:
    ⅝"
  • Wall thickness:
    1/8"
  • Description:
    50' Coil
    (multiple pieces 3' or longer)
  • Cat. no.:
    62995-721
Spécifications
A propos de cet article
Black latex rubber tubing withstands repeated sterilization and has superior tensile strength.
  • Durometer hardness: Shore A, 35
  • Tensile strength: 3500 psi
  • Packaged on 50' reels or in box packs
Black rubber tubing has maximum resistance to aging. Withstands repeated sterilization.
Customers Who Bought This Also Bought