VWR® Latex Rubber Tubing, Black
# de catalogue 62995-721
Fournisseur: VWR International
Spécifications
- Tubing I.D.:⅜"
- Tubing O.D.:⅝"
- Wall thickness:1/8"
- Description:50' Coil
(multiple pieces 3' or longer) - Cat. no.:62995-721
Spécifications
A propos de cet article
Black latex rubber tubing withstands repeated sterilization and has superior tensile strength.
- Durometer hardness: Shore A, 35
- Tensile strength: 3500 psi
- Packaged on 50' reels or in box packs
Black rubber tubing has maximum resistance to aging. Withstands repeated sterilization.
Customers Who Bought This Also Bought