Erlenmeyer Shake Flasks with Three Baffles, Wheaton
# de catalogue 62401-595
Fournisseur: DWK Life Sciences
Small Business Enterprise
Spécifications
- Capacity:2000 ml
- Cat. no.:62401-595
- Supplier No.:353266
Spécifications
A propos de cet article
Designed for use on either rotary or reciprocating shakers.
- Autoclavable
- Specially designed to maximize aeration for improved growth
- Pinched neck design to reduce splashing on flasks with four baffles
Feature a marking spot on the side of the flask and a break-resistant lip, which accommodates a cotton plug or stopper. Made of wheaton 33® low-extractable borosilicate glass, which meets ASTM Type I Class A and USP Type I standards.
Customers Who Bought This Also Bought