VWR® Stopwatch, ¹/₁₀ sec.
# de catalogue 62387-205
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- Cat. no.:62387-205
- Diameter:5.5 cm (2⁵/₃₂")
Spécifications
A propos de cet article
Compact, rugged, and simple to use, with time-out and safety reset features.
- Timing to 0.1 second, plus time-out
- Color-coded, push-button control
- Shock-protected, pin-lever movement
- ABS plastic case
All push-buttons are logically color-coded to minimize timing error and ensure efficient use — green for start, red for stop, and black for return to zero position.
Instant flyback function resets and starts immediately.
The 30-second dial is graduated in tenths of a second, and a 0 to 15 minute register is color-contrasted for easy reading.
Customers Who Bought This Also Bought